missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDCA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513564
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513564 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513564 Supplier Invitrogen™ Supplier No. PA513564
Only null left

Rabbit Polyclonal Antibody

CDCA3 Polyclonal Antibody for Western Blot, IHC (P), Flow

F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt].

Specifications

Antigen CDCA3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CDCA3
Gene Accession No. Q99618, Q99M54
Gene Alias 2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1
Gene Symbols CDCA3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 41-69 amino acids from the N-terminal region of human CDCA3.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14793, 83461
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less