missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC27 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine
Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC3, also known as CDC27, a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), is a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. APC3 contains a tetratricopeptide repeat (TPR) region and interacts with mitotic checkpoint proteins including Mad2, p55CDC and BUBR1.
Specifications
Specifications
| Antigen | CDC27 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | CDC27 |
| Gene Accession No. | P30260 |
| Gene Alias | APC3, CDC27Hs, D0S1430E, D17S978E, HNUC |
| Gene Symbols | CDC27 |
| Host Species | Rabbit |
| Show More |