missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDC25B Recombinant Rabbit Monoclonal Antibody (23GB3425)
GREENER_CHOICE

Catalog No. MA552552
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552552 100 μL
MA552553 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552552 Supplier Invitrogen™ Supplier No. MA552552
Only null left

Rabbit Recombinant Monoclonal Antibody

CDC25B Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Tyrosine protein phosphatase which functions as a dosage-dependent inducer of mitotic progression. Required for G2/M phases of the cell cycle progression and abscission during cytokinesis in a ECT2-dependent manner. Directly dephosphorylates CDK1 and stimulates its kinase activity. The three isoforms seem to have a different level of activity.

Specifications

Antigen CDC25B
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3425
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDC25B
Gene Accession No. P30305, P30306
Gene Alias AI604853; CDC25B; CDC25HU2; Cdc25m2; cell division cycle 25B; dual specificity phosphatase Cdc25B; EC 3.1.3.48; M-phase inducer phosphatase 2; MPIP2
Gene Symbols CDC25B
Host Species Rabbit
Immunogen A synthetic peptide of human Cdc25B (175-250AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12531, 994
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less