missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD99 Recombinant Rabbit Monoclonal Antibody (23GB3395)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553386
Description
CD99 Recombinant Monoclonal Antibody for Western Blot, Flow
CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.Chemical Identifiers
| CD99 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CD99 | |
| 1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2 | |
| CD99 | |
| Primary | |
| Affinity chromatography |
| 23GB3395 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14209 | |
| 4267 | |
| A synthetic peptide of human CD99 (15-70AA). | |
| Antibody |
Specifications
| CD99 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CD99 | |
| 1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4267 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 23GB3395 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14209 | |
| CD99 | |
| A synthetic peptide of human CD99 (15-70AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |