missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD99 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Specifications

Antigen CD99
Concentration 0.057 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01673244
View Documents Promotion Details
Invitrogen™
PA137370
1 mL N/A
Only null left
 
PIPA532337
GREENER_CHOICE
View Documents Promotion Details
Invitrogen™
PA532337
500 μL N/A
Only null left
 
Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is pancreas or Ewing's sarcoma.

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.
Specifications

Specifications

CD99
-20°C, Avoid Freeze/Thaw Cycles
Polyclonal
Liquid
IgG
Human
CD99
1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2
CD99
Primary
Antigen affinity chromatography
0.057 mg/mL
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
RUO
PBS with 1% BSA and 0.08% sodium azide; pH 7.6
P14209
4267
Recombinant full length human CD99 protein.
Antibody
Videos
SDS
Documents

Documents