missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, Trihydrate, Crystal, Reagent, ACS, 98.5-102%, Spectrum™ Chemical

Catalog No. 18605555
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605555 50 kg Amber Glass Bottle
18605548 25 g Amber Glass Bottle
18605549 125 g Amber Glass Bottle
18605550 500 g Amber Glass Bottle
18605552 2.5 kg Amber Glass Bottle
18605551 6 x 500 g Amber Glass Bottle
18605553 4 x 2.5 kg Amber Glass Bottle
18605554 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18605555 Supplier Spectrum Chemical Mfg Cor Supplier No. P129550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1295, 14459-95-1, K4Fe(CN)6·3H2O

Spectrum™ Chemical Potassium Ferrocyanide, Trihydrate, Crystal, Reagent, ACS is an inorganic compound that is a salt with a light yellow tint. Mainly used as an anti-caking agent in both road and table salt. It has several other niche uses and within the laboratory to determine potassium permanganate concentration levels. As an ACS grade Reagent, Spectrum Chemical manufactured compounds are used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 50 kg
Percent Purity 98.5 to 102%
Grade ACS/Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Ferrocyanide, Trihydrate, Crystal