missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD95 (APO-1/Fas) Monoclonal Antibody (DX2), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ H084T03Y06A
Description
CD95, also known as Fas or APO-1, is a 46 kDa transmembrane glycoprotein that functions as a cell death receptor in the tumor necrosis factor receptor (TNFR) superfamily. It is expressed by a wide range of hematopoietic and non-hematopoietic cells, including monocytes, neutrophils, lymphocytes, and fibroblasts. CD95 also exists in a soluble form weighing approximately 26 kDa. The interaction of CD95 with its ligand, FasL, induces apoptosis, playing a crucial role in peripheral tolerance and anti-viral immune responses. Upon stimulation, CD95 leads to the aggregation of its intracellular Fas-associated death domains (FADD), formation of the death-inducing signaling complex (DISC), and activation of caspases. This cascade results in apoptosis. CD95 is also involved in activating NF-kappaB, MAPK3/ERK1, and MAPK8/JNK, and transduces proliferative signals in normal diploid fibroblasts and T cells. There are at least eight alternatively spliced transcript variants encoding seven distinct isoforms of CD95. Isoforms lacking the transmembrane domain may negatively regulate apoptosis mediated by the full-length isoform. The Fas/Fas ligand system is implicated in several human diseases, including AIDS, hepatitis, and cancer, highlighting its importance in immune regulation and disease pathology.Specifications
| CD95 (APO-1/Fas) | |
| Monoclonal | |
| 0.4 μg/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| P25445 | |
| FAS | |
| Affinity chromatography | |
| RUO | |
| 355, 486469 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| DX2 | |
| NovaFluor Yellow 700 | |
| FAS | |
| AI196731; ALPS1A; APO1; APO-1; Apo1 antigen; Apo-1 antigen; APO-1 cell surface antigen; Apo-1 Fas; apoptosis antigen 1; apoptosis signaling receptor FAS; apoptosis-mediating surface antigen FAS; APT1; CD95; CD95 antigen; FAS; Fas (TNF receptor superfamily member 6); Fas (TNF receptor superfamily member); Fas (TNF receptor superfamily, member 6); Fas AMA; Fas antigen; Fas antigen (ATP1); Fas cell surface death receptor; Fas receptor; FAS1; FASLG receptor; FASTM; lpr; lymphoproliferation; OTTHUMP00000020045; OTTHUMP00000020046; OTTHUMP00000020051; OTTHUMP00000059646; TNF receptor superfamily member 6; TNFR6; Tnfrsf6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 6; tumor necrosis factor receptor superfamily member 26; Tumor necrosis factor receptor superfamily, member 6 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 κ |