missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD94 Monoclonal Antibody (HP-3D9), APC, eBioscience™
GREENER_CHOICE

Catalog No. 50245687
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50245687 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50245687 Supplier Invitrogen™ Supplier No. 17509442

Mouse Monoclonal Antibody

Description: The HP-3D9 monoclonal antibody reacts with human CD94, a 70-kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is expressed by NK cells, NKT cells, as well as subsets of CD8+ T cells and gamma delta T cells. This molecule is present as a heterodimer with members of the NKG2 family of receptors on the cell surface. CD94 dimerization with NKG2A results in an inhibitory receptor that suppresses NK cell function. In contrast, CD94 association with NKG2C and NKG2E leads to formation of an activating receptor complex that binds DAP12, an adapter protein that contains an ITAM motif. The CD94/NKG2 receptor complex binds the non-classical MHC Class Ib molecule HLA-E and plays important roles in adhesion and NK cell activation. Crossblocking studies indicate that HP-3D9 and DX22 recognize the same epitope. Applications Reported: This HP-3D9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HP-3D9 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Natural killer (NK) cells, like CD94, are a distinct lineage of lymphocytes that mediate cytotoxic activity and secrete cytokines upon immune stimulation. Several genes of the C-type lectin superfamily, including members of the NKG2 family, are expressed by NK cells and may be involved in the regulation of NK cell function. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. Three transcript variants encoding two different isoforms have been found for this gene.

Specifications

Antigen CD94
Applications Flow Cytometry
Classification Monoclonal
Clone HP-3D9
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene KLRD1
Gene Accession No. Q13241, Q38HS3
Gene Alias Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor
Gene Symbols KLRD1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3824, 611360
Target Species Human, Canine
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less