Learn More
Invitrogen™ CD94 Monoclonal Antibody (DX22), PE, eBioscience™

Description
Description: The DX22 monoclonal antibody reacts with human CD94, a 70 kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is present as a heterodimer with NKG2 on the surface. CD94 is expressed by NK cells, a subset of gd T cells, and NKT cells and plays an important role in adhesion and activation of NK cell lineage. Applications Reported: The DX22 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DX22 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD94 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | DX22 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | KLRD1 |
| Gene Accession No. | Q13241 |
| Gene Alias | Cd94; CD94 antigen; CD94 antigen (located within the rat natural killer gene complex); killer cell lectin like receptor D1; Killer cell lectin-like receptor subfamily D member 1; killer cell lectin-like receptor subfamily D, member 1; killer cell lectin-like receptor, subfamily D, member 1; KLRD1; KP43; natural killer cell receptor; natural killer cells antigen CD94; NK cell receptor |
| Show More |