missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD8b Monoclonal Antibody (SIDI8BEE), PE, eBioscience™, Invitrogen™

Catalog No. 12527341
Encompass
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12527341 25 Tests
12527342 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12527341 Supplier Invitrogen™ Supplier No. 12527341

Mouse Monoclonal Antibody

Description: The SIDI8BEE monoclonal antibody reacts with human and rhesus macaque CD8 beta. The CD8b chain associates with the CD8a chain to form the CD8ab heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. The CD8 receptor complex binds to MHC class I and plays a role in T cell development and activation of mature T cells. Long been thought to exist either as a heterodimer of the alpha and beta chain or as a homodimer of alpha chains, beta chains have now been shown to homodimerize as well. SIDI8BEE can detect the CD8 beta chain in both the CD8ab heterodimer and the CD8bb homodimer. SIDI8BEE is cross-reactive to human and rhesus macaque CD8b. Other non-human primate species have not been tested. Applications Reported: This SIDI8BEE antibody has been reported for use in flow cytometric analysis. Applications Tested: This SIDI8BEE antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

The CD8B antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 beta chain isoforms. Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described. A pseudogene, also located on chromosome 2, has been identified.

Specifications

Antigen CD8b
Applications Flow Cytometry
Classification Monoclonal
Clone SIDI8BEE
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Cd8b
Gene Accession No. P10966
Gene Alias CD8; CD8 antigen 37 kDa chain; CD8 antigen beta polypeptide; CD8 antigen beta polypeptide precursor; CD8 antigen beta-chain; CD8 antigen, beta chain; CD8 antigen, beta chain 1; CD8 antigen, beta polypeptide 1 (p37); CD8 antigen, beta-chain; CD8 beta; CD8 beta chain; CD8a; CD8alpha; CD8B; CD8b antigen; CD8b molecule; CD8b molecule pseudogene; CD8B1; CD8beta; CD8BP; LEU2; Leu-2; LY3; Ly-3; Ly-C; Lymphocyte antigen 3; Lyt3; Lyt-3; MAL; OX-8 membrane antigen; p32; P37; T lymphocyte surface glycoprotein beta chain; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3
Gene Symbols Cd8b
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 926
Target Species Human, Monkey
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less