Learn More
Invitrogen™ CD79b Recombinant Armenian Hamster Monoclonal Antibody (HM79-16)

Armenian Hamster Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551882
Description
Reacts with an extracellular epitope of CD79b. Preparation: HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster. Hybridoma supernatants were screened for positive reactivity in surface immunofluorescence of WEHI231 and then the specificity to CD79b was tested by immuno-precipitationRecombinant anti-CD79 antibodies were engineered by cloning and sequencing the full-length IG heavy and light chains of an Armenian hamster (Cricetulus migratorius).
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.Chemical Identifiers
| CD79b | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Armenian Hamster | |
| RUO | |
| PBS with 0.02% ProClin 300 | |
| P15530 | |
| 15985 | |
| HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster. | |
| Antibody |
| HM79-16 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG λ | |
| Mouse | |
| Cd79b | |
| AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
| Cd79b | |
| Primary | |
| Protein A |
Specifications
| CD79b | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P15530 | |
| Cd79b | |
| HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG λ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| HM79-16 | |
| Unconjugated | |
| Cd79b | |
| AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7 | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| 15985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |