missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD79b Monoclonal Antibody (HM79-12), FITC
GREENER_CHOICE

Catalog No. 501137437
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137437 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137437 Supplier Invitrogen™ Supplier No. HMCD79B01
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Armenian Hamster Monoclonal Antibody

CD79b Monoclonal Antibody for Flow

CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.

Specifications

Antigen CD79b
Applications Flow Cytometry
Classification Monoclonal
Clone HM79-12
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene Cd79b
Gene Accession No. P15530
Gene Alias AGM6; B29; B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; CD79B antigen; CD79b molecule; Cd79b molecule, immunoglobulin-associated beta; Ig beta; IGB; Igbeta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7
Gene Symbols Cd79b
Host Species Armenian Hamster
Immunogen Mouse CD79b. from CD79α/CD79β heterodimers purified from mouse (BALB/c x NZB)F1 B cell lymphoma cells, WEHI231.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15985
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less