missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD66 Monoclonal Antibody (CD66a-B1.1), Super Bright™ 436, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 62066842
Encompass
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62066842 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62066842 Supplier Invitrogen™ Supplier No. 62066842

Mouse Monoclonal Antibody

The CD66 molecules are 180-200 kDa glycoproteins of carcinoembryonic antigen family. They are present on all blood granulocytes and some tissue macrophages, but are absent from other hematopoietic cells. The expression of CD66 increases significantly on granulocytes upon their activation. CD66a (BGP1, CEACAM1) and CD66d (CGM1, CEACAM3), as well as CEACAM2 and 4, are transmembrane proteins, whereas CD66c (CEAL, NCA, CEACAM6) and CD66e (CEA, CEACAM5), as well as CEACAM7 and 8, are anchored to the plasma membrane by C-terminal glycosylphosphatidylinositol (GPI) lipid moiety.

Specifications

Antigen CD66
Applications Flow Cytometry
Classification Monoclonal
Clone CD66a-B1.1
Concentration 5 μL/Test
Conjugate Super Bright 436
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CEACAM4
Gene Accession No. O75871, P06731, P13688, P40198, P40199
Gene Alias antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; Carcinoembryonic antigen CGM1; carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 1; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 3; Carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; carcinoembryonic antigen-related cell adhesion molecule 5; Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66a antigen; CD66c; CD66d; CD66d antigen; CD66e; CEA; CEACAM1; CEACAM-1; CEACAM3; CEACAM-3; CEACAM4; CEACAM5; CEACAM-5; CEACAM6; CEACAM-6; CEAL; CGM1; CGM7; CGM7_HUMAN; Cluster of Differentiation 66c; meconium antigen 100; NCA; Non-specific crossreacting antigen; nonspecific cross-reacting antigen; Nonspecific cross-reacting antigen (NCA); non-specific cross-reacting antigen W236; nonspecific cross-reacting antigen W236; normal cross-reacting antigen; pan CEACAM; W264; W282
Gene Symbols CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048, 1084, 1089, 4680, 634
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less