missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD64 Monoclonal Antibody (10.1), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1,340.54
Specifications
| Antigen | CD64 |
|---|---|
| Clone | 10.1 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H066T03B08A
|
Invitrogen™
H066T03B08A |
100 Tests |
N/A
|
|
|||||
|
H066T02B08A
|
Invitrogen™
H066T02B08A |
25 Tests |
N/A
|
|
|||||
Description
CD64 Monoclonal antibody specifically detects CD64 in Human samples. It is validated for Flow CytometrySpecifications
| CD64 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AI323638; AV092959; CD64; CD64 antigen; CD64A; Fc fragment of IgG high affinity Ib receptor; Fc fragment of IgG receptor Ia; Fc fragment of IgG, high affinity Ia, receptor (CD64); Fc fragment of IgG, high affinity Ia, receptor for (CD64); Fc fragment of IgG, high affinity Ib, receptor for (CD64); Fc gamma receptor; Fc gamma receptor Ia; Fc receptor, IgG, high affinity I; Fcg1; Fc-gamma receptor 1; Fc-gamma receptor I A1; Fc-gamma RI; Fc-gamma RIA; FcgammaRI; FcgammaRIa; Fcgr1; FCGR1A; Fcgr1b; FcgRI; FCRI; FLJ18345; high affinity IgG Fc receptor, subunit beta; high affinity immunoglobulin gamma Fc receptor I; high-affinity immunoglobulin gamma Fc receptor I; IGFR1; IgG Fc receptor I; IgG high affinity Fc receptor; IGGHAFC; RP11-196G18.2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 10.1 | |
| Flow Cytometry | |
| NovaFluor Blue 660-120S | |
| Mouse | |
| Human | |
| P12314 | |
| 2209 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| FCGR1A |