missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD56 (NCAM) Monoclonal Antibody (TULY56), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1340.54
Specifications
| Antigen | CD56 (NCAM) |
|---|---|
| Clone | TULY56 |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H013T02B08A
|
Invitrogen™
H013T02B08A |
25 Tests |
Each for $604.08
|
|
|||||
|
H013T03B08A
|
Invitrogen™
H013T03B08A |
100 Tests |
Each for $1,340.54
|
|
|||||
Description
CD56 (NCAM) Monoclonal antibody specifically detects CD56 (NCAM) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow CytometrySpecifications
| CD56 (NCAM) | |
| 0.4 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| TULY56 | |
| Flow Cytometry | |
| NovaFluor Blue 660-120S | |
| Mouse | |
| Human, Monkey, Rhesus Macaque | |
| P13591 | |
| 4684, 693789 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ncam1 |