missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD45R Monoclonal Antibody (RA3-6B2), NovaFluor™ Blue 760, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$620.90 - $1,203.42
Specifications
| Antigen | CD45R |
|---|---|
| Clone | RA3-6B2 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
M011T02B11A
|
Invitrogen™
M011T02B11A |
25 μg |
Each for $620.90
|
|
|||||
|
M011T03B11A
|
Invitrogen™
M011T03B11A |
100 μg |
Each for $1,203.42
|
|
|||||
Description
CD45R Monoclonal antibody specifically detects CD45R in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| CD45R | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| RA3-6B2 | |
| Flow Cytometry | |
| NovaFluor Blue 760 | |
| Rat | |
| Human, Mouse | |
| P06800, P08575 | |
| 19264, 5788 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| PTPRC |