missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD42b Monoclonal Antibody (HIP1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Catalog No. 46042941
Encompass
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46042941 25 Tests
46042942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46042941 Supplier Invitrogen™ Supplier No. 46042941

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

CD42b is a platelet activation marker involved in the process of coagulation as an aggregating factor. It interacts with extracellular matrix as well as with adhesion molecules. CD42b expression and activation seems to be regulated by VEGF and PDGF. CD42b composes together with GPIb beta, GPIX and GPV the GPIb-IX-V receptor complex critical in the process of platelet-rich thrombus formation by tethering the platelet to a thrombogenic surface. CD42b binds to von Willebrand factor (VWF) exposed at a site of vascular injury, as well as to thrombin, coagulation factors XI and XII, high molecular wight kininogen, TSP-1, integrin Mac-1 and P-selectin. The extracellular domain of CD42b by its interactions also contributes to metastasis. Further, CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. Several polymorphisms and mutations have been described in the CD42b, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease.

Specifications

Antigen CD42b
Applications Flow Cytometry
Classification Monoclonal
Clone HIP1
Concentration 5 μL/Test
Conjugate PerCP-eFluor 710
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Gp1ba
Gene Accession No. P07359
Gene Alias Antigen CD42b-alpha; BDPLT1; BDPLT3; BSS; CD42a; CD42B; CD42b-alpha; DBPLT3; Glycocalicin; glycoprotein 1b, alpha polypeptide; glycoprotein Ib (platelet), alpha polypeptide; glycoprotein Ib platelet alpha subunit; glycoprotein Ib platelet subunit alpha; glycoprotein Ibalpha; GP1B; GP1BA; GPIB; GPIb alpha; GP-Ib alpha; GPIbA; GPIbalpha; GPIX; GP-IX; MGC34595; mutant platelet membrane glycoprotein Ib-alpha; platelet glycoprotein Ib alpha chain; platelet glycoprotein Ib alpha polypeptide; Platelet GPIX; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha; VWDP
Gene Symbols Gp1ba
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2811
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less