missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 Monoclonal Antibody (5C3), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$262.00 - $580.00
Specifications
| Antigen | CD40 |
|---|---|
| Clone | 5C3 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H027T03R06A
|
Invitrogen™
H027T03R06A |
100 Tests |
Each for $580.00
|
|
|||||
|
H027T02R06A
|
Invitrogen™
H027T02R06A |
25 Tests |
Each for $262.00
|
|
|||||
Description
CD40 Monoclonal antibody specifically detects CD40 in Human samples. It is validated for Flow CytometrySpecifications
| CD40 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| P25942 | |
| 958 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 5C3 | |
| Flow Cytometry | |
| NovaFluor Red 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD40 |