missing translation for 'onlineSavingsMsg'
Learn More

CD4 Monoclonal Antibody (GK1.5), NovaFluor™ Blue 725, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$604.08 - $1170.85

Specifications

Antigen CD4
Clone GK1.5
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M001T03B10A
missing translation for 'viewDocuments'
Invitrogen™
M001T03B10A
100 μg
Each for $1,170.85
Only null left
 
M001T02B10A
missing translation for 'viewDocuments'
Invitrogen™
M001T02B10A
25 μg
Each for $604.08
Only null left
 
Description

Description

CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD4
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen
IgG2b κ
Affinity chromatography
Antibody
GK1.5
Flow Cytometry
NovaFluor Blue 725
Rat
Mouse
P06332
12504
Primary
4°C, store in dark, DO NOT FREEZE!
CD4
Videos
SDS
Documents

Documents