missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Monoclonal Antibody (HIT2), Brilliant Violet™ 480, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$607.48 - $1,286.23
Specifications
| Antigen | CD38 |
|---|---|
| Clone | HIT2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
41-403-8942
|
Invitrogen™
414038942 |
100 Tests |
N/A
|
|
|||||
|
41-403-8941
|
Invitrogen™
414038941 |
25 Tests |
N/A
|
|
|||||
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow CytometrySpecifications
| CD38 | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADPRC 1; ADPRC1; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; Cd38; CD38 antigen; CD38 antigen (ADP-ribosyl cyclase / cyclic ADP-ribose hydrolase); CD38 antigen (p45); CD38 antigen p45; CD38 molecule; CD38H; Cd38-rs1; cluster of differentiation 38; cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NAD+ nucleosidase; NIM-R5 antigen; T10 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| HIT2 | |
| Flow Cytometry | |
| Brilliant Violet 480 | |
| Mouse | |
| Human | |
| P28907 | |
| 952 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD38 |