missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1340.54
Specifications
| Antigen | CD274 (PD-L1, B7-H1) |
|---|---|
| Clone | MIH1 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H089T03Y06A
|
Invitrogen™
H089T03Y06A |
100 Tests |
Each for $1,340.54
|
|
|||||
|
H089T02Y06A
|
Invitrogen™
H089T02Y06A |
25 Tests |
Each for $604.08
|
|
|||||
Description
CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Human samples. It is validated for Flow CytometrySpecifications
| CD274 (PD-L1, B7-H1) | |
| 0.8 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MIH1 | |
| Flow Cytometry | |
| NovaFluor Yellow 700 | |
| Mouse | |
| Human | |
| Q9NZQ7 | |
| 29126 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Cd274 |