missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), NovaFluor™ Red 700, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ H089T03R03A

 View more versions of this product

Catalog No. H089T03R03A


Only null left

Description

Description

Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Specifications

Chemical Identifiers

CD274 (PD-L1, B7-H1)
0.6 μg/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
IgG1 κ
Affinity chromatography
Antibody
MIH1
Flow Cytometry
NovaFluor Red 700
Mouse
Human
Q9NZQ7
29126
Primary
4°C, store in dark, DO NOT FREEZE!
Cd274

Specifications

CD274 (PD-L1, B7-H1)
Monoclonal
0.6 μg/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
Q9NZQ7
Cd274
Affinity chromatography
RUO
29126
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MIH1
NovaFluor Red 700
Cd274
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions