missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH1), NovaFluor™ Red 685, eBioscience™, Invitrogen™
Description
Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Specifications
Specifications
| Antigen | CD274 (PD-L1, B7-H1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MIH1 |
| Concentration | 0.8 μg/Test |
| Conjugate | NovaFluor Red 685 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Cd274 |
| Gene Accession No. | Q9NZQ7 |
| Gene Alias | A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 |
| Show More |