missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD268 (BAFF Receptor) Monoclonal Antibody (8A7), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$620.90 - $1377.83
Specifications
| Antigen | CD268 (BAFF Receptor) |
|---|---|
| Clone | 8A7 |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H078T03B09A
|
Invitrogen™
H078T03B09A |
100 Tests |
Each for $1,377.83
|
|
|||||
|
H078T02B09A
|
Invitrogen™
H078T02B09A |
25 Tests |
Each for $620.90
|
|
|||||
Description
CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow CytometrySpecifications
| CD268 (BAFF Receptor) | |
| 0.4 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| 8A7 | |
| Flow Cytometry | |
| NovaFluor Blue 690 | |
| Mouse | |
| Human | |
| Q96RJ3 | |
| 115650 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| TNFRSF13C |