missing translation for 'onlineSavingsMsg'
Learn More

CD268 (BAFF Receptor) Monoclonal Antibody (8A7), NovaFluor™ Blue 510, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$604.08 - $1,340.54

Specifications

Antigen CD268 (BAFF Receptor)
Clone 8A7
Concentration 0.4 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H078T02B01A
View Documents
Invitrogen™
H078T02B01A
25 Tests N/A
Only null left
 
H078T03B01A
View Documents
Invitrogen™
H078T03B01A
100 Tests N/A
Only null left
 
Description

Description

CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD268 (BAFF Receptor)
0.4 μg/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C
IgG2a κ
Affinity chromatography
Antibody
8A7
Flow Cytometry
NovaFluor Blue 510
Mouse
Human
Q96RJ3
115650
Primary
4°C, store in dark, DO NOT FREEZE!
TNFRSF13C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'