missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD267 (TACI) Monoclonal Antibody (ebio8F10-3), APC, eBioscience™, Invitrogen™

Catalog No. 5015259
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5015259 50 μg
5015260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5015259 Supplier Invitrogen™ Supplier No. 17594281
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

Description: The monoclonal antibody eBio8F10.3 recognizes TACI (Transmembrane Activator and Calcium-modulator and CAML Interactor) also known as CD267. TACI is a type III membrane protein belonging to the TNF receptor superfamily. The ligand for TACI has been shown to be BAFF (BLyS) and APRIL. TACI is expressed on the surface of a subset of maturing splenic B cells of the transitional type termed T2. Genetic studies have shown a role for TACI in both stimulation and inhibition of B cell proliferation. This actual role for TACI in the mouse has still not been determined. Recently human TACI has been shown to have repressive activities which inhibit BAFF-R function. Applications Reported: This ebio8F10-3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ebio8F10-3 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.

Specifications

Antigen CD267 (TACI)
Applications Flow Cytometry
Classification Monoclonal
Clone ebio8F10-3
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TNFRSF13B
Gene Accession No. Q9ET35
Gene Alias 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B
Gene Symbols TNFRSF13B
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57916
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only.