missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD265 (RANK) Monoclonal Antibody (R12-31), PE, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 12661282

 View more versions of this product

Catalog No. 501122369


Only null left

Description

Description

Description: The R12-31 monoclonal antibody reacts with mouse RANK (Receptor activator of NF-kappaB), a recently cloned member of the TNFR superfamily with no significant homology to other members of this family. RANK ligand (RANKL/TRANCE/ OPGL) binds to RANK on dendritic cells, upregulates the expression of anti-apoptotic protein BcL-XL suggesting a role in dendritic cell survival. The cytoplasmic domain of RANK interacts with TRAF2, TRAF5 and TRAF6. Overexpression of RANK activates NF-kappaB and c-Jun-terminal kinase (JNK) pathways. Recent studies have shown that RANK interaction with TRAF6 activates NF-kappaB, whereas JNK activation is mediated through binding of RANK to TRAF2. Applications Reported: The R12-31 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R12-31 antibody has been tested by flow cytometric analysis of mouse RANK-transfected cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
Specifications

Chemical Identifiers

CD265 (RANK)
0.2 mg/mL
Monoclonal
Liquid
RUO
O35305
IgG2a κ
Affinity chromatography
Antibody
R12-31
Flow Cytometry
PE
Rat
Mouse
21934
Primary
4°C, store in dark, DO NOT FREEZE!

Specifications

CD265 (RANK)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O35305
TNFRSF11A
Affinity chromatography
RUO
21934
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
R12-31
PE
TNFRSF11A
CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Rat
100 μg
Primary
Mouse
Antibody
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.