missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Monoclonal Antibody (PC61.5), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$604.08 - $1170.85
Specifications
| Antigen | CD25 |
|---|---|
| Clone | PC61.5 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M060T03B06A
|
Invitrogen™
M060T03B06A |
100 μg |
Each for $1,170.85
|
|
|||||
|
M060T02B06A
|
Invitrogen™
M060T02B06A |
25 μg |
Each for $604.08
|
|
|||||
Description
CD25 Monoclonal antibody specifically detects CD25 in Mouse samples. It is validated for Flow CytometrySpecifications
| CD25 | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| ADA; ADA1; Adenosine aminohydrolase; adenosine deaminase; CD25; CD25 antigen; cytokine receptor; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; il-2 receptor alpha; IL-2 Receptor alpha chain; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; IL2R; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2RA; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor; interleukin 2 receptor alpha chain; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha; interleukin-2 receptor alpha chain; interleukin-2 receptor alpha subunit; interleukin-2 receptor beta chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
| IgG1 λ | |
| Affinity chromatography | |
| Antibody |
| PC61.5 | |
| Flow Cytometry | |
| NovaFluor Blue 610-70S | |
| Rat | |
| Mouse | |
| P01590 | |
| 16184 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL2RA |