Learn More
Invitrogen™ CD209a Monoclonal Antibody (LWC06), APC, eBioscience™, Invitrogen™

Description
Description: The LWC06 antibody was generated by immunization with the recombit extracellular region of mouse CIRE/DC-SIGN (CD209). CIRE/DC-SIGN was identified by its expression on CD8 alpha- dendritic cells and plasmacytoid predendritic cells, and is the closest homologue of human DC-SIGN. Human DC-SIGN was originally identified in human placenta for its ability to bind the HIV envelope protein gp120 in a CD4-independent manner. CIRE/DC-SIGN is a 33 kDa type II transmembrane C-type lectin protein. It contains a C-terminal, extracellular, Carbohydrate Recognition Domain (CRD) that is predicted to bind mannose and other carbohydrates in a calcium dependent manner. It has been postulated that CIRE/DC-SIGN may play a role in T-dendritic cell interactions through binding with members of the ICAM family. CIRE/DC-SIGN is differentially expressed by sub-populations of dendritic cells and preliminary data suggest that its expression varies depending on the activation state of the host. CIRE/DC-SIGN is down-regulated in spleen-derived dendritic cell cultures supplemented with GM-CSF. While human DC-SIGN is predomitly expressed in dendritic cells, CIRE/DC-SIGN mRNA has also been detected in B cells. The LWC06 monoclonal antibody does not cross-react with the closely related SIGNR1, SIGNR2, SIGNR3 or SIGNR4. CD209 protein is sensitive to collagenase treatment. Applications Reported: This LWC06 antibody has been reported for use in flow cytometric analysis.
Specifications
Specifications
| Antigen | CD209a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | LWC06 |
| Concentration | 0.2 mg/mL |
| Conjugate | APC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Cd209a |
| Gene Accession No. | Q91ZX1 |
| Gene Alias | CD209; CD209 antigen; CD209 antigen-like protein A; CD209 molecule; Cd209a; CD209a antigen; CD209a molecule; Cd209d; CD209d antigen; CDSIGN; Cire; CLEC4L; Clec4m; C-type lectin domain family 4 member L; C-type lectin domain family 4, member L; C-type lectin domain family 4, member M; Dcsign; DC-SIGN; DC-SIGN1; Dc-signr; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin; dendritic cell-specific ICAM-3-grabbing non-integrin 1; dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin; dendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbing non-integrin; HIV gpl20-binding protein; MGC129965; MGC130443; RGD1561104; SIGN-R1; SIGNR5 |
| Show More |
For Research Use Only.