missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD202b (TIE2) Monoclonal Antibody (33.1), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 50245552
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50245552 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50245552 Supplier Invitrogen™ Supplier No. 14202982
Season of Savings
View all promotions

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This 33.1 monoclonal antibody reacts with human, mouse, rabbit, rat, pig, and zebrafish CD202b. Also known as Tie-2, this molecule is a member of the tyrosine kinase receptor family. CD202b is expressed on endothelial and a subset of hematopoietic cells and is believed to play a role in both angiogenesis and hematopoiesis during development of the mouse embryo. In fetal liver and adult bone marrow, Tie-2 is expressed by a subpopulation of hematopoietic progenitor cells characterized as Lineage markers-, c-Kit+, Sca1+ cells. Long-term multilineage repopulating cells were detected in Tie-2+, Lineage-, c-Kit+, Sca1+ cells but not in Tie-2-, Lineage-, c-Kit+, Sca1+ cells. Applications Reported: This 33.1 antibody has been reported for use in western blotting. Applications Tested: This 33.1 antibody has been tested by immunoblotting of reduced cell lysate prepared from the bEND3 cell line at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells. TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.

Specifications

Antigen CD202b (TIE2)
Applications Western Blot
Classification Monoclonal
Clone 33.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TEK
Gene Accession No. Q02763, Q02858
Gene Alias AA517024; Angiopoietin-1 receptor; CD202B; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; endothelium-specific receptor tyrosine kinase 2; endothelium-specific receptor tyrosine kinase Tie-2; hTIE2; Hyk; mTIE2; p140 TEK; protein-tyrosine kinase Tie2; STK1; Tek; TEK receptor tyrosine kinase; TEK tyrosine kinase, endothelial; TEKT1; Tektin-1; Tie2; tie-2; tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor Tie-2; VMCM; VMCM1
Gene Symbols TEK
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100356675, 21687, 396729, 7010, 89804
Target Species Human, Mouse, Rat, Rabbit, Pig
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.