Learn More
Invitrogen™ Human CD200 ELISA Kit
Description
The Human CD200 ELISA quantitates Hu CD200 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu CD200. Principle of the method The Human CD200 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P41217 |
| Assay Range | 24.58 to 6000 pg/mL |
| Assay Sensitivity | 20 pg/mL |
| Calibration Range | 20 pg/mL to 6000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Product Type | ELISA |
| Protein Family | Other Proteins |
| Sample Type | Plasma, Serum, Supernatant |
| Specificity | This ELISA antibody pair recognizes human CD200. Other species not determined yet. |
| Show More |