missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD19 Monoclonal Antibody (J3-129), PerCP-eFluor™ 710, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ 46019741

 View more versions of this product

Catalog No. 5015931


Only null left

Description

Description

Description: This J3-129 monoclonal antibody reacts with human CD19, a 95-kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development, excluding terminally differentiated plasma cells. However, follicular dendritic cells express CD19. CD21, CD81, Leu13, MHC class II, and CD19 together form a multi-molecular complex that associates with the B cell receptor. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and B cell proliferation. Applications Reported: This J3-129 antibody has been reported for use in flow cytometric analysis. Applications Tested: This J3-129 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.

CD19 is a member of the immunoglobulin superfamily, characterized by two Ig-like domains, and is expressed on B cells throughout all stages of development, excluding terminally differentiated plasma cells. It is also expressed on follicular dendritic cells and has been observed on myeloid leukemia cells, particularly those of monocytic lineage. CD19 is considered the earliest and broadest B cell-restricted antigen, and its expression is found in all B cell precursor leukemias. CD19 forms a multimolecular complex with CD21, CD81, Leu13, MHC class II, and the B cell receptor (BCR), playing a crucial role in B cell signaling. As a signal-amplifying coreceptor for the BCR, CD19 lowers the threshold for antigen receptor-dependent stimulation, allowing B cells to respond specifically and sensitively to various antigens through low-affinity antigen receptors. Signaling through CD19 induces tyrosine phosphorylation, calcium flux, and proliferation of B cells. Beyond its role as a BCR coreceptor, CD19 can also signal independently of BCR co-ligation, serving as a central regulatory component upon which multiple signaling pathways converge. This makes CD19 an important functional regulator of both normal and malignant B cell proliferation. Mutations in the CD19 gene can result in hypogammaglobulinemia, a condition characterized by low levels of immunoglobulins, while CD19 overexpression can lead to B cell hyperactivity. CD19 is expressed on 100% of peripheral B cells, as defined by the expression of kappa or lambda light chains, underscoring its significance in B cell function and immune regulation.
Specifications

Chemical Identifiers

CD19
5 μL/Test
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12
Cd19
Primary
4°C, store in dark, DO NOT FREEZE!
Cd19
J3-129
Flow Cytometry
PerCP-eFluor 710
Mouse
Human
P15391
930
IgG1 κ
Affinity chromatography
Antibody

Specifications

CD19
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P15391
Cd19
Affinity chromatography
RUO
930
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
J3-129
PerCP-eFluor 710
Cd19
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12
Mouse
25 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.