missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD177 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100712 Supplier Invitrogen™ Supplier No. PA5100712
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CD177.

CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).

Specifications

Antigen CD177
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CD177
Gene Accession No. Q8N6Q3
Gene Alias 1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181
Gene Symbols CD177
Host Species Rabbit
Immunogen A synthesized peptide derived from human CD177(Accession Q8N6Q3), corresponding to amino acid residues L298-A348.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499099, 57126
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less