missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD177 Monoclonal Antibody (C11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566047
Description
CD177 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregnancy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).Specifications
| CD177 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8N6Q3 | |
| CD177 | |
| Recombinant Neutrophil Specific Antigen 1 (NB1) (Cys133-Ser301). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C11 | |
| Unconjugated | |
| CD177 | |
| 1190003K14Rik; CD177; CD177 antigen; CD177 molecule; cell surface receptor; HNA2A; HNA-2a; Human neutrophil alloantigen 2a; NB1; NB1 glycoprotein; NB1 GP; Pdp3; Polycythemia rubra vera protein 1; PRV1; PRV-1; RGD1562941; UNQ595/PRO1181 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100519153, 57126 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |