Learn More
Invitrogen™ CD163 Monoclonal Antibody (TNKUPJ), Brilliant Ultra Violet™ 661, eBioscience™
Description
This TNKUPJ monoclonal antibody recognizes mouse CD163. This TNKUPJ antibody will detect CD163 on fixed and permeabilized cells allowing for staining of the intracellular pool of this receptor. Although CD163 is relatively stable to collagenase digestion, aggressive tissue dissociation protocols might potentially decrease the amount of surface CD163. In these cases intracellular detection is recommended. Applications Tested This TNKUPJ antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Blocking Buffers When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information. Light sensitivity This tandem dye is sensitive to photo-induced oxidation. Please protec...
Specifications
Specifications
| Antigen | CD163 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TNKUPJ |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Ultraviolet 661 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD163 |
| Gene Accession No. | Q2VLH6 |
| Gene Alias | CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 |
| Show More |