missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Monoclonal Antibody (MAC 2-158), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$604.08 - $1340.54
Specifications
| Antigen | CD163 |
|---|---|
| Clone | MAC 2-158 |
| Concentration | 0.4 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H088T02B09A
|
Invitrogen™
H088T02B09A |
25 Tests |
Each for $604.08
|
|
|||||
|
H088T03B09A
|
Invitrogen™
H088T03B09A |
100 Tests |
Each for $1,340.54
|
|
|||||
Description
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow CytometrySpecifications
| CD163 | |
| 0.4 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MAC 2-158 | |
| Flow Cytometry | |
| NovaFluor Blue 690 | |
| Mouse | |
| Human | |
| Q86VB7 | |
| 9332 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| CD163 |