missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine, 98.5-101.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Heavy Metals | 1.5e-05 |
| Loss on Drying | 0.002 |
| Packaging | Poly Pail |
| Specific Rotation | +12.6° to +14.0° |
| Molecular Weight (g/mol) | 155.15 |
| Quantity | 5 kg |
| Percent Purity | 98.5 to 101.5% |
| Grade | Reagent |
| Chemical Name or Material | L-Histidine |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.