missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD162 (PSGL-1) Monoclonal Antibody (4RA10), PE-eFluor™ 610, eBioscience™

Catalog No. 61162182
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61162182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61162182 Supplier Invitrogen™ Supplier No. 61162182
Season of Savings
View all promotions

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

This 4RA10 monoclonal antibody reacts with mouse CD162, which is also known as P-selectin glycoprotein ligand-1 (PSGL-1). This clone has been successfully used in flow cytometry and has been reported to block binding of CD162 to CD62P and CD62L. Applications Reported: This 4RA10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4RA10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PE-eFluor 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red). Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-57) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/comp...

CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.

Specifications

Antigen CD162 (PSGL-1)
Applications Flow Cytometry
Classification Monoclonal
Clone 4RA10
Concentration 0.2 mg/mL
Conjugate PE-eFluor 610
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene SELPLG
Gene Accession No. Q62170
Gene Alias CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG
Gene Symbols SELPLG
Host Species Rat
Immunogen Mouse PSGL-1 human IgG1 fusion protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20345
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less