missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD160 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596968 Supplier Invitrogen™ Supplier No. PA596968
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: MLLEPGRGCC ALAILLAIVD IQSGGCINIT SSASQEGTRL NLICTVWHKK EEAEGFVVFL CKDRSGDCSP ETSLKQLRLK RDPGIDGVGE ISSQLMFTIS QVTPLHSGTY QCCARSQKSG IRLQGHFFSI LFTETGNYTV TGLKQRQHLE FSHNEGTLSS GFLQEKVWVM LVTSLVALQA L; Positive Samples: Jurkat, HT-29, U-937, Raji, Mouse spleen, Mouse small intestine, Mouse thymus, Rat thymus; Cellular Location: Cell membrane, GPI-anchor, Lipid-anchor.

CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. It is involved in cell proliferation, cell surface receptor signaling pathways, cellular defense response, and regulation of immune response.

Specifications

Antigen CD160
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD160
Gene Accession No. O88875, O95971
Gene Alias AU045688; BY55; CD160; CD160 antigen; CD160 antigen, soluble form; CD160 molecule; CD160 transmembrane isoform; CD160-delta Ig; natural killer cell BY55; natural killer cell receptor; natural killer cell receptor BY55; natural killer cell receptor, immunoglobulin superfamily member; NK1; NK28; RGD1563598
Gene Symbols CD160
Host Species Rabbit
Immunogen Recombinant protein (or fragment.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11126, 502585, 54215
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less