Learn More
Invitrogen™ CD150 Monoclonal Antibody (mShad150), eFluor™ 450, eBioscience™
Description
Description: The mShad150 monoclonal antibody reacts with mouse CD150, an ~70 kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T cells, in particular Th1, and B cells; this expression is rapidly upregulated upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. The mShad150 antibody has been reported to also stain the CD150+CD244-CD48-CD41- population of pluripotent hematopoietic stem cells. Applications Reported: This mShad150 antibody has been reported for use in flow cytometric analysis. Applications Tested: This mShad150 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorocho...
Spécifications
Spécifications
| Antigen | CD150 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | mShad150 |
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 450 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Slamf1 |
| Gene Accession No. | Q9QUM4 |
| Gene Alias | 4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; Slamf1 |
| Afficher plus de résultats |