missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD140a (PDGFRA) Monoclonal Antibody (APA5), Brilliant Ultra Violet™ 661, eBioscience™
GREENER_CHOICE

Catalog No. 376140182
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
376140182 100 μg
376140180 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 376140182 Supplier Invitrogen™ Supplier No. 376140182
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rat Monoclonal Antibody

The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the alpha chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues. Applications Tested: This APA5 antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Blocking BuffersWhen using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information.Light sensitivityTh...

PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.

Specifications

Antigen CD140a (PDGFRA)
Applications Flow Cytometry
Classification Monoclonal
Clone APA5
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 661
Formulation PBS with BSA and 0.09% sodium azide, pH 7.2
Gene PDGFRA
Gene Accession No. P26618
Gene Alias AI115593, alpha platelet-derived growth factor receptor, Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha), alpha-type platelet-derived growth factor receptor, APDGFR, CD140 antigen-like family member A, CD140A, CD140a antigen, GAS9, MGC74795, OTTHUMP00000218656, PDGF A-chain, PDGF alpha chain, PDGF Receptor alpha, PDGF subunit A, PDGF-1, PDGFACE, PDGFR2, Pdgfr-2, PDGFRA, pdgfr-a, PDGFRA/BCR fusion, PDGF-R-alpha, PDGFR-alpha, platelet derived growth factor receptor alpha, platelet derived growth factor receptor, alpha polypeptide, platelet-derived growth factor alpha receptor, Platelet-derived growth factor receptor 2, platelet-derived growth factor receptor alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein, RHEPDGFRA
Gene Symbols PDGFRA
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18595
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less