missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD130 Recombinant Rat Monoclonal Antibody (KGP130)

Rat Recombinant Monoclonal Antibody

$639.17 - $1,120.94

Specifications

Antigen CD130
Clone KGP130
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
740182T
View Documents
Invitrogen™
740182T
100 μL N/A
Only null left
 
740182TXS
View Documents
Invitrogen™
740182TXS
20 μL N/A
Only null left
 
Description

Description

Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications

Specifications

CD130
1 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rat
RUO
PBS with 0.09% sodium azide; pH 7.4
Q00560
16195
Primary
Affinity chromatography
KGP130
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG2a κ
Mouse
IL6ST
5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha
IL6ST
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'