missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD11d Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149793
Description
CD11d Polyclonal Antibody for Western Blot, ICC/IF, IHC
Integrin alpha-D/beta-2 is a receptor for ICAM3 and VCAM1. May play a role in the atherosclerotic process such as clearing lipoproteins from plaques and in phagocytosis of blood-borne pathogens, particulate matter, and senescent erythrocytes from the blood.Specifications
| CD11d | |
| Polyclonal | |
| Unconjugated | |
| ITGAD | |
| ADB2; beta-2 integrin alphaD subunit; CD11 antigen-like family member D; CD11d; FLJ39841; integrin alpha X (Cd11c); integrin alpha-D; integrin subunit alpha D; integrin, alpha D; ITGAD; Itgax; Leukointegrin alpha D | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3681, 381924, 64350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q13349, Q3V0T4, Q9QYE7 | |
| ITGAD | |
| Recombinant Integrin Alpha D (ITGaD) (Phe886-Glu1106). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |