Learn More
CD107b (LAMP-2) Monoclonal Antibody (eBioH4B4 (H4B4)), FITC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 11107841
Description
Description: The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease. Applications Reported: This eBioH4B4 (H4B4) antibody has been reported for use in intracellular staining followed by flow cytometric analysis. It has also been reported for use in surface staining in a flow cytometric based degranulation assay. Applications Tested: This eBioH4B4 (H4B4) antibody has been pre-titrated and tested by intracellular staining and flow cytometric analysis of Jurkat cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Lysosomes are membrane-bound vacuoles which play a critical role in cellular metabolism. These organelles participate in endocytosis, the synthesis and assembly of acid hydrolases, act as sites for digestion of foreign materials and for specialized autolytic cellular processes. Membrane-associated glycoproteins have an important role in normal lysosomal function. Prior to posttranslational modification, Lysosome Associated Membrane Protein 2 (LAMP 2) is an ∽45 kDa polypeptide. Mature, functional LAMP 2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of ∽100-110 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail.Specifications
| CD107b (LAMP-2) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
| P13473 | |
| LAMP2 | |
| Affinity chromatography | |
| RUO | |
| 3920 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| eBioH4B4 (H4B4) | |
| FITC | |
| LAMP2 | |
| RP23-193O17.2, CD107b, Lamp-2, Lamp-2a, Lamp-2b, Lamp-2c, Mac3 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
For Research Use Only.