missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD107a (LAMP-1) Monoclonal Antibody (eBioH4A3), Brilliant Violet™ 650, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$607.48 - $1286.23
Specifications
| Antigen | CD107a (LAMP-1) |
|---|---|
| Clone | eBioH4A3 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
41-610-7941
|
Invitrogen™
416107941 |
25 Tests |
Each for $607.48
|
|
|||||
|
41-610-7942
|
Invitrogen™
416107942 |
100 Tests |
Each for $1,286.23
|
|
|||||
Description
CD107a (LAMP-1) Monoclonal antibody specifically detects CD107a (LAMP-1) in Human samples. It is validated for Flow CytometrySpecifications
| CD107a (LAMP-1) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| 120 kDa lysosomal membrane glycoprotein; AI196048; CD107 antigen-like family member A; CD107a; I79_011073; Lamp I; LAMP1; LAMP-1; LAMPA; LGP120; LGP-120; LGPA; LGP-A; lysosomal associated membrane protein 1; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; lysosomal membrane glycoprotein A; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| eBioH4A3 | |
| Flow Cytometry | |
| Brilliant Violet 650 | |
| Mouse | |
| Human | |
| P11279 | |
| 3916 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| LAMP1 |