Learn More
Invitrogen™ CD106 (VCAM-1) Monoclonal Antibody (429), FITC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Supplier: Invitrogen™ 11106181
Description
Description: The 429 monoclonal antibody reacts with mouse CD106 (Vascular Cell Adhesion Molecule-1, VCAM-1), a 110 kDa transmembrane glycoprotein expressed by myeloid lineage and bone marrow stromal cells. Endothelial cells constitutively express low levels of CD106 and upregulate it upon cytokine stimulation. CD106 binds to integrin alpha4beta1 (VLA-4, CD49d/CD29) and Integrin a4b7 (LPAM-1) and these interactions in the bone marrow and thymus are important for early lymphocyte and myeloid development. Cytokine-mediated upregulation of CD106 on endothelial cells suggests a role for this antigen in the inflammatory response. Applications Reported: The 429 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 429 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.Chemical Identifiers
| CD106 (VCAM-1) | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| VCAM1 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| VCAM1 |
| 429 | |
| Flow Cytometry | |
| FITC | |
| Rat | |
| Mouse | |
| P29533 | |
| 22329 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Specifications
| CD106 (VCAM-1) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P29533 | |
| VCAM1 | |
| Affinity chromatography | |
| RUO | |
| 22329 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 429 | |
| FITC | |
| VCAM1 | |
| adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
| Rat | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
For Research Use Only.