missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD OptiCHO™ Medium

Catalog No. p-7180304
Encompass
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50591551 1 L
12681029 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50591551 Supplier Gibco™ Supplier No. 12681011
Only null left

CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO's legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advancedCHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.

When to Use CD OptiCHO™ Medium

  • You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
  • Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
  • Your CHO cell line is 'finicky,' i.e. hard to grow or adapt

When to Consider Another Gibco™ Medium

Consider CD FortiCHO™ medium when:

  • The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
  • Maximum batch culture cell densities and protein titers are needed
  • You have time to adapt cells into a new medium in an effort to maximize titers
  • Cell health at harvest is a priority for downstream processing

Consider CD CHO medium when:

  • Your GS CHO cells are recommended to be grown in CD CHO medium
  • Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)

Consider CD DG44 medium when:

  • You use parental DG44 or DXB11 CHO cells (dhfr-)

Dry Media Formulations

Gibco™ liquid media are available in an AGT™ (Advanced Granulation Technology) dry media format that is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. AGT™ media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their liquid media counterparts.

Design Your Own Media Specific to Your Needs

Whether it's a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs.

Specifications

Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type CD OptiCHO Medium
Serum Level Serum-free
With Additives Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red
Product Line Gibco
Quantity 1 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Bottle
Included Antibiotics No antibiotics
Species Hamster
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Suspension Cell Culture
Show More Show Less