missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD EfficientFeed™ C AGT™ Nutrient Supplement

Catalog No. A1327504
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1327504 1 L
A1327505 10 L
A1327501 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1327504 Supplier Gibco™ Supplier No. A1327504

CD EfficientFeed™ C AGT™ Nutrient Supplement

GIBCO™ CD EfficientFeed™ C AGT™ nutrient supplement is a dry format single part supplement for fed batch addition to the culture of multiple cell lines. This product is designed to work with GS CHO cell lines as a single feed without the need for additional GS supplements. CD EfficientFeed™ C AGT™ nutritient supplement is an animal origin-free, chemically defined formulation that contains no proteins, hydrolysates, or components of incompletely defined composition.

The AGT™ format is designed for large scale manufacturing and ease of use.

  • Up to 2.5 – 6 Fold increases versus batch culture – higher concentration of key components
  • Higher productivity in GS CHO and other selected cell lines
  • Avoid issues associated with the use of hydrolysates and other alternatives while gaining superior performance
  • Simplify scale-up by leveraging the value of AGT™ simple reconstitution
  • Ready for larger scale use with convenient packaging options (1 L, 10 L, and 100 L)
  • No components of animal origin, minimizing the opportunity for introduction of adventitious agents and facilitating the regulatory process

Which EfficientFeed™ Nutrient Supplement to Choose?

Consider CD EfficientFeed™ C AGT™ supplement when:

  • You want higher productivity in GS CHO and other selected cell lines
  • You are currently using CD FortiCHO™ as your base cell growth medium
  • You are using a base medium with greater than 6 g/L glucose or a hydrolysate at day 0

Consider CHO CD EfficientFeed™ A supplement (Cat. No. A1023401) when:

  • You are currently using CD OptiCHO™ as your base cell growth medium

Consider CHO CD EfficientFeed™ B supplement (Cat. No. A1024001) when:

  • You are currently using CD CHO as your base cell growth medium

Consider the CHO CD Efficient Feed™ Kit (Cat. No. A1024101) when:

  • You are using a base medium with less than 6 g/L glucose and no hydrolysate at day 0

Increased Titers

CHO CD EfficientFeed™ supplements have been shown to fold increases in titers versus batch culture, and are an excellent starting point for process development. Furthermore, it does not require addition of a GS Supplement when used in CHO cell lines expressing endogenous glutamine synthetase.

Simple Scale-Up and Cost Savings

The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CD EfficientFeed™ C AGT™ supplement is offered in 1 L, 10 L and 100 L packaging that allows scale-up as a seamless process, as well as allow for cost savings when purchasing production level volumes.

AGT™ Format Ease of Use

CD EfficientFeed™ C AGT™ Nutrient Supplement delivers dry format scale-up convenience as single part AGT dry format. For more details, consult the supporting documents provided below.

For research or further manufacturing use. Not for use in diagnostic procedures.

Specifications

Content And Storage Store in the dark at 2°C to 8°C. Stable for 24 months when properly stored.
Culture Environment CO2
Antibiotic Spectrum No Antibiotics
Cell Line CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Culture Type Suspension Cell Culture
Form AGT
Product Type Media Supplement
Serum Level Serum-Free
Sterility Non-sterile
With Additives Sodium Bicarbonate
Without Additives No Glutamine, No Phenol Red, No Sodium Pyruvate, No Insulin
Endotoxin Level Low
Format Securitainer Capped Vial
Purity or Quality Grade Cell Culture Grade, Qualified for Further Manufacturing, Research Grade
Quantity 1 L
Remarks Chemically Defined Nutrient Feeding Supplement
Shipping Condition Wet Ice
Source Animal origin-free
Tested For Solubility
Show More Show Less

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.