missing translation for 'onlineSavingsMsg'
Learn More

CD CHO Medium

Catalog No. 10743001
Change view
Click to view available options
Quantity:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10743001 10 L
10743002 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10743001 Supplier Thermo Fisher Scientific Supplier No. 10743001

Specifications

Cell Line CHO
Cell Type Ovarian Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Concentration 1 X
Form Liquid
Product Type CD CHO Medium
Serum Level Serum-free
Sterility Sterile-filtered
With Additives Sodium Pyruvate, Sodium Bicarbonate, Non Essential Amino Acids (NEAA)
Without Additives No Glutamine, No Phenol Red, No Thymidine, No Hypoxanthine
Product Line Gibco
Quantity 10 L
Shipping Condition Room Temperature
Endotoxin Level Low
Format Universal Bag
Included Antibiotics No antibiotics
Purity or Quality Grade Cell Culture Grade
Concentration Ratio 1 :1
Content And Storage Store at 2-8°C in the dark.
Culture Environment CO2
Culture Type Suspension Cell Culture
Show More Show Less