missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD-1000 Medium

Catalog No. 215206
Change view
Click to view available options
Quantity:
500 g
1 L
10 kg
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
215206 500 g
215207 10 kg
215204 1 L
215205 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 215206 Supplier Gibco™ Supplier No. 215206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A chemically defined (CD), animal origin-free (AOF) basal medium that is ideally suited for further hydrolysate or chemically defined supplementation

CD-1000 Medium can be used in various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications.

Specifications

Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Form Liquid
Product Type Cell Culture Media
For Use With (Application) Various culture vessels in batch, fed-batch, and profusion processes for both research and manufacturing applications
Quantity 500 g
Source Animal origin-free, chemically defined